Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
RiboPrinter® System Disposable, Hygiena™
Supplier: Qualicon Diagnostics
ASY2004 ASY2077 KIT2030 ASY2074 ASY2028 ASY2078 KIT2032 ESA RP D12416018 KIT2033 KIT2034 KIT2036 KIT2029
10014-838EA 121.76 USD InStock
10014-838 10014-834 10014-846 10014-868 10014-858 10014-836 10014-864 10014-842 10014-866 10014-844 10014-870 10014-860 10014-840
RiboPrinter® System Disposable, Hygiena™
Microbiology Test Systems Microorganism Tests
Riboprinter® system accurately identifies contaminants and track their source at the strain level.
  • Database of about 7000 patterns, covering more than 180 genera and 1400 species
  • Highly precise differentiation among strains of the same species
  • Fast, automated answers with minimal labor required
  • Reliable and easy to use
Bacteria are found throughout nature, their presence in certain critical environments, such as sterile manufacturing or food processing plants, poses a constant threat
Unlike other systems just analyzing the region that encodes the 16S rRNA sequence, the RiboPrinter® System investigates the regions encoding the 5S, 16S and 23S sequences, as well as the spacer regions and flanking genes on either side. This rich depth of information is what allows highly precise differentiation among strains of the same species, even those with the same 16S sequence.
The RiboPrinter® System technology investigates a much larger portion of the bacterial genome than other bacterial identification systems can. Also this system helps to accurately determine the source of infections, enabling the hospital to take quick action in replacing the incubators.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty