Would you like to log in again?
Biopsy Cassettes, Leica
Supplier: Leica
3802463 3802453 3802465 3802459 3802461 3802457 3802455
10014-962CS 248.52 USD InStock
10014-962 10014-964 10015-138 10014-960 10014-966 10014-968 10015-140
Biopsy Cassettes, Leica
Embedding Cassettes Biopsy Processing Cassettes
Biopsy Cassettes are designed to allow processing of the smallest specimens without the need to use foam inserts or lens paper.
  • Rigid construction
  • Eliminates need for foam inserts or lens paper
  • Complete fluid exchange and thorough paraffin infiltration
These rigidly constructed cassettes are made of solvent- and acid-resistant plastic featuring a textured writing surface on three sides. Biopsy cassettes also feature 1x1mm square holes to permit complete fluid exchange and thorough paraffin infiltration. Cassettes also feature a "stay-shut" closure design and a 50° writing surface.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty