Would you like to log in again?
Mounting Media, Leica
Supplier: Leica
3801721 3801705 3801743 3801731 3801732 3801731 3801720
10014-984CS 350.34 USD InStock
10014-984 10014-974 10014-986 10014-976 10014-980 89130-990 10014-978
Mounting Media, Leica
Stains Microscopy and staining reagents
Acrytol® is a rapid-drying mounting medium which enables slides to be screened immediately.
  • Low viscosity allows for easy flowing
  • Rapid-drying
  • Xylene-based mounting medium
  • Added antioxidant inhibits stain fading and prevents formation of annual rings
It has a low viscosity, allowing the medium to flow easily
Micromount® is a rapid-drying, xylene-based mounting medium. Its low viscosity facilitates coverslipping and prevents air bubbles from forming. An antioxidant is added to inhibit stain fading and to prevent the formation of annual rings. Micromount® sets up quickly, allowing slide preparations to be handled immediately after mounting. Micromount® is soluble in xylene, toluene, and most xylene substitutes.
Sub-X® Mounting Medium is a quick-drying, permanent mountant formulated to be compatible with a Sub-X® Clearing Agent and all xylene substitutes on the market. It is toluene-based and may also be used on coverslip preparations cleared with xylene or toluene. Its' low viscosity provides an easy flow, bubble-free medium suitable for manual as well as automated coverslipping. Sub-X® contains an antioxidant to inhibit fading of stains.
It also prevents air bubbles from becoming trapped. Acrytol® contains an antioxidant to inhibit stain fading and to prevent the formulation of annual rings. A thin coating of Acrytol® is all that is needed to adhere a coverglass to a microscope slide. It is soluble in xylene, toluene, and most xylene substitutes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty