Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Scalpel Blades, Leica
Supplier: Leica
3802160 3802186 3802182 3802181 3802187
10014-996BX 122.02 USD InStock
10014-996 10014-992 10014-994 10014-990 10014-988
Scalpel Blades, Leica
Scalpels and Scalpel Assemblies Scalpel Blades
The Feather Autopsy, Trimming, and Dissection range is internationally renowned for its sharpness and versatility.
  • Stainless steel or carbon steel construction
  • Scalpel tips are available in a variety of formats
  • Supplied sterile or nonsterile
  • Excellent feel and dexterity
The #22 feather brand scalpel blades are stainless steel, which provide a durable, precision edge
A diverse range of scalpel blades and handles for use in the pathology laboratory and mortuary are also available. The #22 and #60 scalpels are available in the traditional pointed tip format or in the #22B and #60B rounded tip or bullnose format for enhanced user safety. These scalpels are made of the finest carbon steel and are supplied nonsterile.
The Safeshield disposable scalpel features a stainless steel Feather blade protected by a plastic safety shield that can be locked in place or moved. The protective shield slides easily back and forth across the handle and locks into either an open or closed position to expose or cover the blade. The ergonomically designed handle offers ease of use and user comfort while the sliding protective shield ensures an added measure of safety.
They are packaged individually and are sterile to maximize their applications. Designed to be used with a #4 Stainless Steel contoured handle which accentuates proper feel and dexterity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty