Would you like to log in again?
SelecTech Define Staining System, Leica
Supplier: Leica
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Surgipath®
3802918 3802916 3801571 3803595 3801575 3801619 3801615 3803591 3803598 3801606 3801570
10015-088CS 855.18 USD InStock
10015-088 10015-078 75810-358 75839-344 75810-354 10015-080 75810-352 10015-086 10015-076 10015-084 75810-360
SelecTech Define Staining System, Leica
Stains Histology Stains
SelecTech is a hematoxylin and eosin staining system consisting of Hematoxylin 560, Blue Buffer 8, Define, Alcoholic Eosin Y 515 and Eosin Trichrome 515. SelecTech Ready-To-Use Define aq is available in concentrates as well as ready-to-use formulas. Define is a refinement product designed to create crisp, nuclear detail and cosistency.
SelecTech standardizes the reagents and stains used in the hematoxylin and eosin staining procedure.
The built-in quality assurance parameters assure consistent results. The reagents and stains in SelecTech are designed to complement each other to produce a crisp, well delineated hematoxylin and eosin stain.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty