Would you like to log in again?
ST Infinity H&E Staining System, Leica
Supplier: Leica
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3801698
10015-090CS 356.09 USD InStock
10015-090
ST Infinity H&E Staining System, Leica
Stains Histology Stains
All stains and reagents are optimized for use with Leica ST5010 and Leica ST5020, resulting in high-quality H&E stains and consistent quality from the first stained slide to the last.
  • Consistently high quality
  • Enables high throughput of slides before stains expire
  • Expediency allows all components to be loaded simultaneously
This system enables a high throughput of slides before the stains and reagents expire. The time saving, as all stains and reagents exhaust at the same rate, allows all components of the kit to be loaded on and off the instrument at the same time.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty