Would you like to log in again?
Snowcoat® Microscope Slides, Leica
Supplier: Leica
Snowcoat®
3800340 3800310 3800320 3800305 3800299
10015-188CS 519.82 USD InStock
10015-188 10015-182 10015-180 10015-186 10015-184
Snowcoat® Microscope Slides, Leica
Slides Microscope Slides
Snowcoat® Slides feature a colored marking area printed on the most wettable surface of the slide for easy marking and color-coded identification.
  • Colored marking area
  • Color-coded identification
  • Easy to mark
  • Printed surface facilitates microscope focusing
The printed surface facilitates microscope focusing and identifies the best side to use, even by touch if necessary.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty