Would you like to log in again?
Bead Ruptor Pre-Filled Bead Tubes, 15 ml, Omni International
Supplier: Omni International
19-6158
10015-218EA 468.94 USD InStock
10015-218
Bead Ruptor Pre-Filled Bead Tubes, 15 ml, Omni International
Tubes Centrifuge Tubes
Bead beating is the most effective and versatile lysis method available.
This 15 ml polypropylene tube with leakproof screw cap is vacuum-chamber tested to 95 kPa. Pre-filled with lysing matrix. Non sterile tubes are designed for use on the Omni International Bead Ruptor. Compatible with solvents, alcohols, chlorinated and aromatic hydrocarbons (except xylene), and ketones.
Omni International offers ultimate flexibility with a range of bead beating tubes and lysing matrices to support efficient homogenization of a wide range of samples. Bead Ruptor bead beating tubes are pre-filled with an optimized amount of lysing matrix to provide efficient, consistent, and high yield sample homogenates.
Ordering information: Unit includes 50 bead beating tubes pre-filled with lysing matrix.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty