Would you like to log in again?
Resprep® Tools for ASE® Systems, Restek
Supplier: Restek
26181
10015-286PK 220.59 USD InStock
10015-286
Resprep® Tools for ASE® Systems, Restek
Extractors Automatic Extractor Systems
Resprep® tools are specialized to simplify routine chores and suitable for ASE 100/200/300.
  • Use to insert filter in extraction cell or O-ring in cell cap
  • Fits all extraction cells, except 1 mL size
Handle is easy to grip, and various attachments alllow for compatibility with extraction cells of all sizes.
Delivery information: Includes Resprep tool handle and filter insertion attachments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty