Would you like to log in again?
Sodium Sulfate (Bulk Adsorbent), Restek
Supplier: Restek
26204
10015-308EA 277.24 USD InStock
10015-308
Sodium Sulfate (Bulk Adsorbent), Restek
Spill Control Materials
Anhydrous sodium sulfate is the most common drying agent used to remove moisture from sample extracts.
  • Ideal for removing water from sample extracts
  • Activate by heating to 400°C for four hours before use
  • Packaged in recloseable 5 kg buckets
We package Restek's 10-60 mesh material in recloseable buckets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty