Would you like to log in again?
Extraction Cell Bodies for ASE® Systems, Restek
Supplier: Restek
25994 26114 25995 26098 26116 26112 26178 25996 25993 26110 26176 26132
10015-420EA 147.8 USD InStock
10015-420 10015-388 10015-386 10015-430 10015-424 10015-428 10015-390 10015-280 10015-426 10015-262 10015-384 10015-282
Extraction Cell Bodies for ASE® Systems, Restek
Chromatography Supplies Chromatography Spare Parts
Accelerated solvent extraction is a common technique for fast and reliable extraction of organic materials from solid matrices, using EPA SW-846 Method 3545, Pressurized Fluid Extraction (PFE).
  • Cell bodies are serialized for easy sample identification
  • Smooth inner surfaces for easier cleaning
Restek offers a wide range of replacement parts to keep your extraction system running smoothly. All parts are economically priced to save you money and are designed to meet or exceed the performance of the original manufacturer’s parts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty