Would you like to log in again?
Diatomaceous Earth (Bulk Adsorbent), Restek
Supplier: Restek
26033
10015-398EA 389.35 USD InStock
10015-398
Diatomaceous Earth (Bulk Adsorbent), Restek
Spill Control Materials
Diatomaceous earth is used as a filter aid to improve extraction efficiency of densely packed soils, such as clays.
  • Improves extraction efficiency
  • Adsorbs moisture from samples
By mixing the sample with diatomaceous earth, recoveries can be improved and excess moisture can be absorbed. Packaged in a convenient 1 kg quantity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty