Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

20 mm Filters for ASE® 200 Extraction Cells, Restek

Supplier: Restek

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

26119 26118 26190
10015-432PK 50.39 USD InStock
10015-432 10015-434 10015-302
20 mm Filters for ASE® 200 Extraction Cells, Restek
Filters In-Line Filters
For use with ASE® 200 extraction cells.

  • Cellulose or glass fiber construction
  • Fit 11 ml, 22 ml, and 33 ml cells
  • Cellulose filters available in economical 1000 packs

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty