Would you like to log in again?
Tryptic Soy Broth (TSB), Hardy Diagnostics
Supplier: Hardy Diagnostics
K85
10015-554PK 75.26 USD InStock
10015-554
Tryptic Soy Broth (TSB), Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Tryptic Soy Broth (TSB), filtered to remove dead bacteria.
Tryptic Soy Broth (TSB) is recommended for use as a general purpose medium for the isolation and cultivation of a wide variety of bacteria and fungi.
Ordering information: Minimum order of 5 packs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty