Would you like to log in again?
Tryptone Broth 2%, Hardy Diagnostics
Supplier: Hardy Diagnostics
R40 R40
10015-568PK 126.7 USD InStock
10015-568 CA10015-568
Tryptone Broth 2%, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the performance of the Indole test.
For the performance of the Indole test. Use with Kovacs Reagent.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty