Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Transparent Bath Tanks For Use With Heating Immersion Circulator To +100 °C, JULABO USA, Inc.
Supplier: Julabo
9901319 9901309 9901305 9901327
10015-824EA 582.17 USD InStock
10015-824 10015-822 10015-828 10015-826
Transparent Bath Tanks For Use With Heating Immersion Circulator To +100 °C, JULABO USA, Inc.
Baths Water Baths, Unstirred
Transparent Plexiglass bath tanks.
  • Clear Plexiglass tanks
  • Rim with integrated handles
  • Suitable for use with CORIO C/CD heating immersion circulators
Transparent bath tanks for sample incubation in conjunction with heating immersion circulators. Tank volumes available in 9, 19 and 27 l sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty