Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Baths, Stainless Steel, JULABO
Supplier: Julabo
9903427 9903417 9903439 9903433 9903419 9903413 9903405
10015-842EA 4098.86 USD InStock
10015-842 10015-832 10015-840 10015-830 10015-836 10015-834 10015-838
Baths, Stainless Steel, JULABO
Baths Water Baths, Unstirred
Stainless steel bath tanks for use with heating immersion circulator up to +150 °C.
  • Insulated stainless steel tanks
  • Integrated handles in the rim
  • Suitable for use with CORIO C/CD heating immersion circulators
These tanks are used for sample incubation in conjunction with heating immersion circulators.
Ordering information: Tank volumes available in 5, 13, 17, 19, 27, 33 and 39 L sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty