Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Omni Mixer and Macro Homogenizers, Omni International
Supplier: Omni International
17505 17105 17506 17106
10015-894EA 7848.69 USD InStock
10015-894 CA11011-580 10015-892 CA11011-584
Omni Mixer and Macro Homogenizers, Omni International
Homogenizers Benchtop Homogenizers
Since 1956 the Omni Mixer Homogenizers have been synonymous with laboratory homogenizers.
The Omni Mixer Homogenizer is compatible with stainless steel generator probes ranging in diameter from 7 to 35 mm, and glass, plastic, or stainless steel chambers from 13 to 1360 ml with sealed generator probes or blades. The Omni Macro Homogenizer is compatible with stainless steel generator probes with a 7, 10, 20, 32, 45, and 55 mm diameter, and glass, plastic, or stainless steel chambers from 13 to 1360 ml with sealed generator probes or blades.
The Omni Mixer Homogenizer features a processing range of 0.25 ml to 10 l, while the Omni Macro Homogenizer is scalable to 30 l volumes.
Optional stainless steel Chamber Assemblies are available in 50, 200, or 400 ml sizes. All formats feature a fluted chamber design to facilitate sample flow and mixing, as well as autoclavable chamber and components. Chambers feature a sealing gasket made of Viton®. All chamber sizes are ideal for fibrous tissue and for pre-processing to reduce particle sizes to 1 to 2 μm.
The Omni Mixer and Macro Homogenizer are powerful 600 and 1800 watt homogenizers that are compatible with a wide range of generator probes, blade assemblies, and sealed containers to offer the perfect mix of dependability, versatility, and power. The Omni mixers are ideally suited for homogenizing, blending, and creating emulsions.
Certifications: CE compliant.
Ordering information: Unit includes 600 (Mixer) or 1800 (Macro) watt variable speed motor, motor stand assembly, generator probe adapter, tool kit, and instruction manual. Each unit is also supplied with a three-year motor warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty