Would you like to log in again?
DuPont™ Tyvek® IsoClean® Coveralls with Standard Collar
Supplier: DuPont
Tyvek® IsoClean®
IC181SWHLG00250C IC181SWH4X002500 IC181SWHLG002500 IC181SWH5X002500 IC181SWH7X002500 IC181SWHMD002500 IC181SWH4X00250C IC181SWH2X00250C IC181SWH3X002500 IC181SWH3X0C IC181SWHSM002500 IC181SWHSM002500 IC181SWH2X002500 IC181SWHXL00250C IC181SWHXL002500 IC181SWH8X002500 IC181SWHSM00250C IC181SWHXL002500 IC181SWHSM00250C IC181SWHMD00250C IC181SWH6X002500
80076-748CS 304.43 USD InStock
80076-748 80076-948 76187-472 CA80076-786 80076-744 80076-788 CA80076-744 80076-746 80077-288 10016-066 10016-068 80076-794 80076-750 10016-070 80076-796 80076-752 CA80076-750 80076-786 80076-790 80076-792 10016-064
DuPont™ Tyvek® IsoClean® Coveralls with Standard Collar
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Standard laydown collar
  • Set sleeve design
  • Zipper closure
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls have elastic openings for a tighter fit at wrists and ankles. Collar and closure allow for easy donning and doffing.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Packaging: Clean Processed garments are individually packaged.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty