Would you like to log in again?
DuPont™ ProShield® 60 Coveralls with Respirator Hood
Supplier: DuPont
Pro Shield
NG127SWHSM0025NP NG127SWHLG0025NP NG127SWH4X0025NP NG127SWHMD0025NP NG127SWH3X0025NP NG127SWHXL0025NP NG127SWH5X0025NP NG127SWH6X0025NP NG127SWH7X0025NP NG127SWH2X0025NP
10016-120CS 184.68 USD InStock
10016-120 10016-110 10016-122 10016-112 10016-124 76187-484 10016-114 10016-116 10016-118 10016-108
DuPont™ ProShield® 60 Coveralls with Respirator Hood
Disposable Garments
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Respirator fit hood
  • Elastic wrists and ankles
  • Storm flap
  • Serged seams
ProShield® 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Attached respirator-fit hood with elastic around face opening was designed to cover neck and chin and fit around respirator face mask. Longer zipper extends to chin for complete coverage of neck area. Storm flap of garment material covers zipper closure. Elastic openings at wrists and ankles allows for a tighter fit.
Certifications: Thickness meets ASTM D751.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty