Would you like to log in again?
DuPont™ ProShield® 60 Boot Covers with PVC Sole
Supplier: DuPont
ProShield®
NG456SWHLG010000 NG456SWHSM010000
10016-136CS 246.29 USD InStock
10016-136 10016-140
DuPont™ ProShield® 60 Boot Covers with PVC Sole
Disposable Garments
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • PVC skid-resistant soles
  • Serged seams
  • Elastic top
  • Coverage up to calf
ProShield® 60 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation.
These 18" boot covers provide coverage up to the calf and feature an elastic top and PVC soles for enhanced skid-resistance and durability.
Certifications: Thickness meets ASTM D751.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty