Would you like to log in again?
DuPont™ Tychem® 4000 Coveralls with Standard Hood and Attached Socks, Bound Seams
Supplier: DuPont
Tychem®
SL122BWH6X001200 SL122BWHLG0012BN SL122BWH2X001200 SL122BWH5X001200 SL122BWHMD001200 SL122BWH3X001200 SL122BWHMD001200 SL122BWH3X0012BN SL122BWH2X0012BN SL122BWH4X0012BN SL122BWHXL001200 SL122BWH4X001200 SL122BWH3X001200 SL122BWHXL001200 SL122BWHLG001200 SL122BWH7X001200 SL122BWHXL0012BN SL122BWH4X001200 SL122BWH2X001200 SL122BWHMD0012BN SL122BWHLG001200
10010-154CS 439.95 USD InStock
10010-154 CA10010-164 CA10010-154 10010-164 10010-162 CA10010-156 CA10010-160 10010-158 CA10010-162 10010-156 CA10010-158 10016-472 10016-462 10016-474 10016-464 10016-476 10016-466 10016-478 10016-468 10010-160 10016-470
DuPont™ Tychem® 4000 Coveralls with Standard Hood and Attached Socks, Bound Seams
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration. Coveralls feature an attached hood with elastic around face opening and elastic openings at wrists for a tighter fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Integrated socks are composed of the same garment material.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty