Would you like to log in again?
DuPont™ Tychem® 4000 Coveralls with Respirator Fit Hood and Attached Socks
Supplier: DuPont
SL128TWHXL000600 SL128TWH5X000600 SL128TWH4X000600 SL128TWHMD000600 SL128TWH2X000600 SL128TWHSM000600 SL128TWHLG000600 SL128TWH3X000600
10016-510CS 527.27 USD InStock
10016-510 10016-512 10016-502 10016-514 10016-504 10016-516 10016-506 10016-508
DuPont™ Tychem® 4000 Coveralls with Respirator Fit Hood and Attached Socks
Disposable Garments
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
  • Respirator fit hood
  • Elastic wrists
  • Attached socks with outer boot flaps
  • Storm flap with adhesive closure
  • Taped seams
Tychem® 4000, utilizes Saranex™ 23-P film laminated to Tyvek® fabric to provide a rugged and durable fabric that resists abrasion. Tychem® 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications include chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Attached respirator-fit hood features elastic around face opening, which is designed to cover neck and chin and to fit around a respirator face mask. Coveralls also feature a strong zipper for tough, flexible closure. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrists allows for a tighter fit. Integrated socks are composed of the same garment material. Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty