Would you like to log in again?
DuPont™ Tyvek® 400 Boot Covers with FC Skid Resistant Sole
Supplier: DuPont
Tyvek®
TY454SWHLG0100SR TY454SWHLG0100SR
10016-958CS 236.19 USD InStock
10016-958 CA10016-958
DuPont™ Tyvek® 400 Boot Covers with FC Skid Resistant Sole
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Elastic top
  • Coverage up to calf
  • Special FC slip-resistant coating on sole
  • One size fits most
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include: lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Sole material is a special Tyvek® FC skid-resistant coating to provide added slip resistance.
Certifications: Thickness meets ASTM D1777.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty