Would you like to log in again?
Specialty NMR Sample Tubes, Controlled Atmosphere Valve (CAV), New Era™
Supplier: New Era
NECAV5XXX NECAV10XXX
100165-918EA 174.28 USD InStock
100165-918 100165-914
Specialty NMR Sample Tubes, Controlled Atmosphere Valve (CAV), New Era™
Tubes NMR Tubes
Precision thin wall NMR Sample Tubes feature a valve for air/moisture sensitive samples.
  • Made of type 1 class A borosilicate glass
  • Low pressure to five bar
A linear valve system for performing controlled atmosphere experiments by NMR. The concentric valve mechanism consists of a sturdy, precision machined TFE plug with a viton O-ring seal. A glass vacuum adapter is supplied with each system. It is secured to the top of the valve with a press fit sealed by an O-ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty