Would you like to log in again?
NMR Sample Tube Washers, New Era™
Supplier: New Era
NE240
100165-924EA 90.09 USD InStock
100165-924
NMR Sample Tube Washers, New Era™
Tubes NMR Tubes
This basic style washer is an economical, convenient and effective way to clean NMR sample tubes.
  • Tube washers feature a a 24/40 inner joint
  • Reservoir capacity is 8 ml
Insert the washer body into a vacuum flask, invert the sample tube over the central capillary, and secure it in place with a modified tube cap. When a vacuum is applied and the reservoir is filled with solvent, a spray of solvent washes the tube and is collected in the flask. Several solvents may be used in sequence followed by an 'air rinse' to dry the tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty