Would you like to log in again?
NMR Sample Tubes, 3 mm, New Era™
Supplier: New Era
NEH37 NEH38
100165-952EA 15.02 USD InStock
100165-952 100165-936
NMR Sample Tubes, 3 mm, New Era™
Tubes NMR Tubes
Improved design allows for increased sample volume while maintaining adequate wall thickness to help reduce breakage. For use in all 3 mm micro probes.
  • Made of type 1, class A borosilicate glass
  • Tubes are suitable for flame sealing to Pyrex® or equivalent
  • Each tube is capped and marked with the catalog number
Ideal for biological samples, hands-on and high throughput/autosampler use, and samples not requiring special handling. Suitable for all experiments when matched to the application and instrument field strength.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty