Would you like to log in again?
Tip-Off NMR Sample Tubes, New Era™
Supplier: New Era
NEUP5TTO NEHP5TTO
100165-958EA 39.03 USD InStock
100165-958 100165-946
Tip-Off NMR Sample Tubes, New Era™
Tubes NMR Tubes
A 50 mm section of 5 mm Ext.Ø tubing is sealed to a sample tube. The extra tubing allows the tube to be secured in the tip-off adapter with ample length for safe flame sealing. A uniform constriction, with a 2 mm Int.Ø is placed just above the 7" length.
  • Made from type 1, class A borosilicate glass
  • Flame sealable
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty