Would you like to log in again?
Specialty NMR Sample Tubes with Screw Thread, New Era™
Supplier: New Era
NEHL5ST168 NEUL5ST168 NEH10ST178
100165-962EA 51.78 USD InStock
100165-962 100165-960 100165-966
Specialty NMR Sample Tubes with Screw Thread, New Era™
Tubes NMR Tubes
Tubes offer the convenience of a screw cap with the security of a PTFE/silicone seal.
  • Made of type 1 class B borosilicate glass
  • 5 mm Ext.Ø tubes fit all standard 5 mm probes
  • 10 mm Ext.Ø tubes fit all standard 10 mm probes
Each tube includes an open-top cap and a PTFE/silicone septum. A 22 gauge, standard point needle is recommended for a clean puncture. To maintain spinning quality, choose the tube length that places the threads closest to the spinner.
These tubes are ideally suited for glove box preparation of air or moisture sensitive samples. The screw cap provides easy access for kinetic studies. Another proven application has been tracking the evolution of hydrolysis products within bioabsorbable materials.
Ordering information: Optional lengths are available via special order.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty