Would you like to log in again?
Thin Walled Series NMR Sample Tubes, 10 mm, New Era™
Supplier: New Era
NEH108 NEH107 NEL108 NEL107
100165-984EA 31.49 USD InStock
100165-984 100165-970 100165-954 100165-986
Thin Walled Series NMR Sample Tubes, 10 mm, New Era™
Tubes NMR Tubes
All large volume NMR sample tubes are made of type 1, class A borosilicate glass (Pyrex or equivalent), unless otherwise noted.
  • Round bottom is standard
  • Flat bottoms are available upon request
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty