Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AlphaTec® 62-401 Chemical resistant gloves, Ansell

Supplier: Ansell

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AlphaTec®
62401110 62401080 62401100 113916 62401090
10017-172CS 541.3 USD InStock
10017-172 75795-324 10017-170 10017-176 10017-174
AlphaTec® 62-401 Chemical resistant gloves, Ansell
Gloves
Type 62-401 gloves have a double dipped natural rubber coating with wrinkle finish for added grip.

  • High degree of flexibility
  • Waterproof
  • Added grip
  • Thermal insulation in hot and cold extremes

These gloves are comfortable for periods of long use, offer a high level of flexibility, and are waterproof.

For use in construction, glass, public works, and food industries. Uses include handling sharp objects in a messy and/or humid environment.

62-401 gloves deliver thermal insulation in either hot (up to 250 °C/482 °F) or cold (down to −20 °C/−4 °F) conditions.

Certifications: 21 CFR (USA) compliant for food applications. ANSI Cut Level 2. ANSI Abrasion Level 3.

Packaging: Packaged 6 pairs per bag; 8 bags per carton.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty