Would you like to log in again?
Vesta-Syde® SQ st Quaternary Ammonium Disinfectant, STERIS®
Supplier: Steris
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1S22CX 634029
10017-668CS 741.49 USD InStock
10017-668 76303-118
Vesta-Syde® SQ st Quaternary Ammonium Disinfectant, STERIS®
Cleaning Agents Disinfectants
Vesta-Syde® SQ st quaternary ammonium disinfectant is a sterile, liquid concentrate specifically formulated for disinfection of hard surfaces.
  • Flexibility - Excellent fungicidal efficacy can support reduced sporicide use to control mold contamination
  • Convenience - Gamma irradiated and double bagged for easy introduction into your aseptic areas
  • Productivity - Formulated with biodegradable surfactants that provide excellent cleaning ability
  • Availability - Active Ingredients notified according to the Biocide Product Directive and registered with the US EPA
  • Assurance - Each lot documented by a Certificate of Analysis including irradiation and sterility results
At a use-dilution rate of 0.8% v/v (1:128), the Vesta-Syde SQ products kill a broad spectrum of common bacteria and fungi without harming the surface.
The use-dilution can be applied manually or sprayed.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty