Would you like to log in again?
flashBAC™ Baculovirus Expression Systems, Mirus Bio
Supplier: Mirus Bio
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
flashBAC™
MIR6115 MIR6130 MIR6135 MIR6120 MIR6140 MIR6125
10017-678EA 3295.17 USD InStock
10017-678 10017-676 10017-684 10017-682 10017-680 10017-708
flashBAC™ Baculovirus Expression Systems, Mirus Bio
Transfection Reagents
The flashBAC system is a streamlined platform designed for the production of recombinant baculoviruses.
  • Ease-of-use - single-step baculovirus generation in insect cells, no plaque purification required
  • Compatibility - ready to use with most transfer vectors (PDF), including BacMam
  • More protein - high recombinant protein yields
Further nonessential gene deletions in flashBAC ULTRA provide enhanced quality and yield for difficult to express proteins. The insertion of a bacterial artificial chromosome (BAC) at the AcMNPV polyhedrin gene locus allows viral DNA to be maintained and propagated as a circular genome within bacterial cells. The genomic DNA can then be purified and is the flashBAC DNA provided with this kit. To generate recombinant baculovirus using the flashBAC system, insect cells are transfected with TransIT® -Insect Transfection Reagent, flashBAC DNA and a transfer plasmid (pOET1, pOET1C_6xHIS, or pOET6) containing the gene of interest.
Homologous recombination within the insect cells inserts the gene of interest under the control of the polyhedrin promoter, and restores the function of ORF1629 allowing viral DNA to replicate and produce virus particles. The recombinant virus genome replicates to produce baculovirus that can be harvested directly from the culture medium of transfected insect cells. Because the flashBAC System has effectively reduced recombinant baculovirus production to a one-step procedure, it is fully amenable to high throughput and automated production platforms.
Deletion of the essential gene ORF1629 from the Autographa californica nucleopolyhedrovirus (AcMNPV) genome prevents non-recombinant virus from replicating within insect cells, thus eliminating the need to plaque-purify recombinant virus from parental virus. Secreted or membranetargeted recombinant protein yields are greatly increased by the deletion of the chitinase gene (chiA).
Ordering information: flashBAC ULTRA Baulovirus Expression Systems are provided with respective flashBAC DNA and Control Transfer Plasmid (pAcRP23-lacZ). pOET Transfer Plasmids are available separately and designed for use with the flashBAC Baculovirus Expression Systems and TransIT®-Insect Transfection Reagent.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty