Would you like to log in again?
WHEATON® Erlenmeyer Shake Flasks with DuoCap® Closures, DWK Life Sciences
Supplier: DWK Life Sciences
WPFBC2000S WPFBC3000S WPFBC0500S WPFBC1000S
10017-888CS 211.53 USD InStock
10017-888 10017-894 10017-892 10017-890
WHEATON® Erlenmeyer Shake Flasks with DuoCap® Closures, DWK Life Sciences
Flasks Erlenmeyer Flasks
WHEATON® offers both flat bottom and baffled Erlenmeyer flasks.
  • Ideal replacement for glass and other Erlenmeyer/shaker flasks
  • Patented DuoCap® allows sterile air exchange or a leak-resistant seal
  • Minimize oxidation or pH shifts
  • Perform aerobic/anaerobic culture
  • Available with septum cap for pathogen containment
  • Re-autoclavable, reusable
  • Individually packed in easy tear bags
These Erlenmeyer flasks are made of Lexan polycarbonate resin, which offers outstanding mechanical, optical, electrical, and thermal properties. This resin conforms to flame retardant standards and is scratch-resistant. It also meets the requirements of United States Pharmacopeia (USP) XXII Class VI. The flasks are manufactured in a class 10K clean room, and are certified pyrogen-, DNase- and RNase-free. They are packaged particulate-free and wrapped in an easy tear, sterile bag.
Both Erlenmeyer and shake flasks are supplied standard with a patented vented DuoCap®. The DuoCap®, made of medical grade polypropylene, has a 0.22 µm membrane that allows sterile gas exchange when the cap is open. When the cap is closed, the flask becomes leak-resistant. Baffled flasks are ideal for applications that require increased levels of oxygenation and mixing. The flask and DuoCap® can be autoclaved (less than or equal to 10 cycles) and can withstand temperatures up to 130 °C. Each Flask is gamma irradiated to sterility assurance level (SAL) of 10⁻⁶. The flasks are transparent and feature molded graduation markings.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty