Would you like to log in again?
KIMBLE® KIMAX® Pink Colorware Volumetric Flasks, Class A, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE® KIMAX®
28014PNK-250 28014PNK-100 28014PNK-500 28014PNK-1000 28014PNK-25 28014PNK-50
10017-936CS 521.93 USD InStock
10017-936 10017-926 10017-934 10017-932 10017-930 10017-928
KIMBLE® KIMAX® Pink Colorware Volumetric Flasks, Class A, DWK Life Sciences
Flasks Volumetric Flasks
Please join us to support breast cancer prevention, treatment, and research by purchasing Pink Colorware from DWK Life Sciences.
  • Manufactured from low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Pink markings - ideal for color coding
  • Single graduation ring is calibrated "To Contain"
  • Glass stopper included, along with a durable, matte marking area for labeling
  • Replacement stopper: 850100 series
Choose from a wide variety of KIMBLE® KIMAX® Pink “Colorware”, including Low Form Griffin Beakers, Narrow Mouth Erlenmeyer Flasks, Class A Volumetric Flasks, Blood Typing Tubes, and GL45 caps for ease of identification in the lab.
We will donate 20% of the proceeds to the Breast Cancer Research Foundation, the highest rated breast cancer organization in the United States. BCRF is recognized as one of the most financially efficient nonprofits in the country, and we are proud to assist in their support of breast cancer research.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty