Would you like to log in again?
Brady® B30 Series Printer Ribbon R10000 Series
Supplier: Brady
140851 140856 140864 140848 140855 140865 140857 140859 140854 140847 140845 140852 140850 140862 140863 140860 140858 140846 140866 140849
10017-972EA 179.78 USD InStock
10017-972 10017-950 10017-984 10017-940 10017-970 10017-982 10017-958 10017-948 10017-978 10017-956 10017-968 10017-946 10017-954 10017-976 10017-966 10017-944 10017-952 10017-974 10017-942 10017-964
Brady® B30 Series Printer Ribbon R10000 Series
Printer Ribbons
B30 Series Printer Ribbon R1000, available in Single Color, Two Color, or Four Color options.
  • Tough, smear-proof ribbon produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions
  • Self-contained B30 Series cartridges drop straight into the printer with a "click"
  • R10000 series ribbons are based on a resin formulation that offers a high performance image with strong solvent and scratch resistance
"Drop & Click" print ribbons for quick and easy supply changes. Ribbons are tough and smear-proof, and they produce long-lasting printed text. Print can withstand years in industrial areas and outdoor conditions. No calibrating, no threading, no waste, no hassles. Paneled ribbons prevent swapping out ribbons during multicolor print jobs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty