Would you like to log in again?
Mix-n-Stain™ Antibody Labeling Kits, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
92329 92328 92244 92286 92266 92295 92327 92326 92325 92296 92294 92330
75832-530EA 184.48 USD InStock
75832-530 75832-640 75832-826 89411-604 89411-616 75832-828 10018-228 10018-232 75832-824 75832-638 10018-230 89411-610
Mix-n-Stain™ Antibody Labeling Kits, Biotium
Protein Linking and Labeling Systems
Mix-n-Stain™ antibody labeling kits dramatically simplify the process of labeling primary antibodies with fluorescent dyes or biotin
Mix-n-Stain™ Antibody Labeling Kits feature Biotium's line of CF™ dyes, which offer advantages in brighness and/or photostability compared to Alexa Fluor® dyes and other similar commercial dyes. A large selection of CF™ dye colors spanning the visible and near-infrared spectra offers flexibility for labeling primary antibodies with dye colors that are optimal for your instrument detection settings for multi-color detection. See the CF™ Dye Reference Table under the Datasheets tab for more information.
CF™ dye technologies are covered by pending US and international patents. Mix-n-Stain™ and CF™ are trademarks of Biotium, Inc. AlexaFluor® is a registered trademark of Molecular Probes, Inc.
Simply mix your antibody with the buffer and premeasured aliquot of CF™ dye provided in the kit, a step that takes less than 30 seconds of hands-on time. After a 30 minute incubation, your fluorescent antibody conjugate is ready to use, with no purification step. There is no need to calculate how much dye you should use; just select the approptriate kit size based on the ug amount of antibody you wish to label for optimal results. Moreover, the labeling reaction can tolerate the presence of common stabilizers such as sodium azide and low levels of Tris, glycerol, BSA or gelatin. An ultrafiltration column provided in the kit can be used to remove Tris, glycine, or glycerol if the concentration in your antibody solution exceeds the limits specified in the labeling protocol. See the product protocol under the Datasheets tab for more information.
Ordering information: One labeling reaction per kit. Kits are sized according to amount of antibody to be labeled (50-100 ug, 20-50 ug, or 5-20 ug antibody per labeling).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty