Would you like to log in again?
CF® Dye Alkyne, Biotium
Supplier: Biotium
92090 96004 92088 92089 96005 96006 92086 92091 92095 92087 92093
10018-402EA 338.46 USD InStock
10018-402 10018-404 10018-398 75832-768 10018-400 10018-224 75832-764 10018-394 75832-766 10018-396 10018-220
CF® Dye Alkyne, Biotium
Protein Linking and Labeling Systems
CF® Dye alkynes react with azide groups via copper-catalyzed bioorthogonal cycloaddition. Can be used to fluorescently detect or label azide groups on target molecules. CF® Dye alkynes can also be used as building blocks to form fluorescent polymers.
  • Fluorescent detection or labeling azide groups on target molecules
  • Can be used as fluorescent monomers to form dye-labeled polymers
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labeling
CF® Dye alkynes can react with azide groups to form 1,2,3-triazole via 1,3-dipolar Huisgen copper-catalyzed cycloaddition. They can also be used as monomeric building blocks to copolymerize with other monomers to form fluorescent polymers.
CF® Dyes are Biotium's line of next-generation fluorescent dyes that have improved brightness, photostability and water solubility compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty