Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dispensette® S Bottle Top Dispensers

Supplier: Brand

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


Dispensette® S
4600330 4600310 4600210 4600121 4600161 4600331 4600220 4600130 4600351 4600170 4600231 4600360 4600320 4600340 4600341 4600361 4600151 4600321 4600211 4600160 4600171 4600230 4600150 4600240 4600100 4600311 4600140 4600241 4600221 4600131 4600350 4600120 4600101 4600141
10018-944EA 1165.1 USD InStock
10018-944 10018-966 10018-968 10018-946 10018-940 10018-962 10018-964 10018-942 10019-010 10018-948 10019-014 10019-012 10020-964 10020-966 10018-970 10018-972 10018-950 10020-960 10020-962 10018-934 10018-956 10018-978 10018-936 10018-958 10018-974 10018-930 10018-952 10018-976 10018-932 10018-954 10018-938 10019-016 10018-980 10018-960
Dispensette® S Bottle Top Dispensers
Bottle Top Dispensers
Fixed volume or variable volume, analogue-adjustable or digital easy calibration.

  • With or without recirculation valve
  • Autoclavable at 121 °C
  • Three different models of volume adjustment available: Fixed (fixed volume for standard applications; simple calibration adjustment with supplied tool) and Analogue (analogue volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy calibration technique for adjustment in seconds without tools; alteration of factory setting is automatically indicated
  • Easy to calibrate and adjust in order to comply with ISO 9001 and GLP guidelines; alteration of factory setting is automatically indicated

Dispensette® S supports a wide range of applications for the dispensing of aggressive reagents directly from the supply bottle; such as concentrated bases and acids like H₃PO₄, H₂SO₄ (with certain exceptions such as HCl, HNO₃, HF, and so on), saline solutions, and a variety of organic solvents. Parts that are in contact with the liquid are made from borosilicate glass, ceramic, platinum-iridium, ETFE, FEP, PFA, PTFE and PP (discharge tube safety screw cap). Colour code: Red.

* Special fixed volumes: 0.5 to 100 ml (please state when ordering)!

Ordering information: Dispensette® S is supplied with a DE-M marking, performance certificate, telescopic filling tube, recirculation tube (optional), mounting tool and PP adapters.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty