Would you like to log in again?
Micro Clip, Straight, 0.8×5 mm, Roboz
Supplier: Roboz
RS-5420
100188-862EA 116.31 USD InStock
100188-862
Micro Clip, Straight, 0.8×5 mm, Roboz
Forceps
Micro Clip; Straight; 70 Grams Pressure; 0.75 mm Clip Width; 5 mm Jaw Length.
The jaws have fine atraumatic serrations. Please use RS-5410 or RS-5440 clip applying forceps with these clips. Compressing the clips with fingers is very difficult and it may damage them.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty