Would you like to log in again?
Micro Clip, Straight, Strong Curved, 1×6 mm, Roboz
Supplier: Roboz
RS-5433
100188-894EA 137.08 USD InStock
100188-894
Micro Clip, Straight, Strong Curved, 1×6 mm, Roboz
Forceps
Micro Clip; Straight Strong Curve; 70 Grams Pressure; 1 mm Clip Width; 6 mm Jaw Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty