Would you like to log in again?
Johns Hopkins Bulldog Clamps, Roboz Surgical
Supplier: Roboz
RS-7443 RS-7446 RS-7440 RS-7442 RS-7444 RS-7445 RS-7447 RS-7441
100188-872EA 126 USD InStock
100188-872 100188-870 100188-908 100188-918 100188-906 100188-890 100188-876 100188-920
Johns Hopkins Bulldog Clamps, Roboz Surgical
Clamps General Purpose Clamps
The Bulldog clamp is a useful instrument for stopping blood flow to a particular organ of interest
These premium quality bulldog clamps have a rivet in the shank. The jaws can be separated by twisting the shanks sideways around the rivet. This provides easy access to the serrations in the jaws for thorough cleaning, which is very difficult when the jaws don't open wide enough or when the closing force keeps them shut during the cleaning cycle.
One of the most common applications where a bulldog clamp can be used is during coronary surgery where blood flow must be stopped. In general the bulldog clamp, with its many angled varieties and surgical applications, is an essential tool during surgery where blood flow to an organ must be stopped or controlled.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty