Would you like to log in again?
Freer Elevator 7", Double Ended 5 mm, Roboz Surgical
Supplier: Roboz
RS-8820
100188-934EA 120.46 USD InStock
100188-934
Freer Elevator 7", Double Ended 5 mm, Roboz Surgical
Retractors
Freer Elevator; Double Ended; Sharp blade:1.0 mm thick; Blunt blade: 1.5 mm thick; 5 mm Width; 7" Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty