Would you like to log in again?
Forceps, 2.75" (7 cm) Long, 1×2 Teeth, 0.12 mm Teeth With Suture Tying Platform, 0.3 mm Tip Width, Roboz
Supplier: Roboz
RS-5170
100188-936EA 434.77 USD InStock
100188-936
Forceps, 2.75" (7 cm) Long, 1×2 Teeth, 0.12 mm Teeth With Suture Tying Platform, 0.3 mm Tip Width, Roboz
Forceps
Bonn Forceps; Tying Platform; 1x2 Teeth, 0.12 mm Teeth ; 2.75" Length; .3 mm Tip Width
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty