Would you like to log in again?
Dumont #N5 Forceps, Cross Action Inox, Thin Tips Size 0.05×0.01 mm, Biologie Tips, Roboz
Supplier: Roboz
RS-5020
100188-974EA 69.23 USD InStock
100188-974
Dumont #N5 Forceps, Cross Action Inox, Thin Tips Size 0.05×0.01 mm, Biologie Tips, Roboz
Forceps
Dumont Tweezers; Pattern #N5; Cross Action; Thin Tips; .05 x .01 mm Tip Size; 108 mm Length; Material: Inox; Biologie Tips
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty