Would you like to log in again?
Dissecting Micro Scissors, Sharp, Roboz Surgical
Supplier: Roboz
RS-5788
100189-002EA 481.85 USD InStock
100189-002
Dissecting Micro Scissors, Sharp, Roboz Surgical
Scissors
Noyes Micro Dissecting Spring Scissors; Angled On Flat (Angled Up); 12 mm Blade Length; 0.25 mm Tip Width, Sharp/Sharp tips, 4 3/4" Overall Length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty