Would you like to log in again?
Jacobson Mosquito Forceps, 5", Curved, Extra Delicate, 0.9 mm tips, Roboz
Supplier: Roboz
RS-7117
100189-122EA 272.77 USD InStock
100189-122
Jacobson Mosquito Forceps, 5", Curved, Extra Delicate, 0.9 mm tips, Roboz
Forceps
Jacobson Micro Mosquito Forceps; Extra Delicate; Light Weight; Curved; 5" Length
The tip width is 0.9 mm. The jaw width is 1.3 mm at 5 mm from the tip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty