Would you like to log in again?
Tube Occluding Forceps, Straight With Lockguard 7.5", Roboz
Supplier: Roboz
RS-7478
100189-126EA 128.77 USD InStock
100189-126
Tube Occluding Forceps, Straight With Lockguard 7.5", Roboz
Forceps
St
Vincent Tube Occluding Forceps; Narrow Jaws; Straight, Serrated; Lockguard; 7.5" Length. The jaw width is 5.5 mm at 5 mm from the tip
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty