Would you like to log in again?
Thumb Dressing Forceps, 4.5", Serrated Delicate 1.3 mm Tip Width, Roboz
Supplier: Roboz
RS-8120
100189-176EA 60.92 USD InStock
100189-176
Thumb Dressing Forceps, 4.5", Serrated Delicate 1.3 mm Tip Width, Roboz
Forceps
Thumb Dressing Forceps; Serrated; Delicate; 4.5" Length; 1.3 mm Tip Width
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty