Would you like to log in again?
Dissecting Spring Scissors, Vannas, Angled On Flat, Roboz Surgical
Supplier: Roboz
RS-5619
100189-252EA 494.31 USD InStock
100189-252
Dissecting Spring Scissors, Vannas, Angled On Flat, Roboz Surgical
Scissors
Vannas Micro Dissecting Spring Scissors; Angled On Flat (Angles Up When Laid Down); 3 mm Cutting Edge; 0.15 mm Tip Width; 3" Overall LengthBlades angled on flat make it easier to do horizontal cutting from above.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty